missing translation for 'onlineSavingsMsg'
Learn More

(R,R)-2,2'-Isopropylidenebis(4-phenyl-2-oxazoline) 96.0+%, TCI America™

Catalog No. D28235G
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D28235G 5 g
D2823250MG 250 mg
D28231G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. D28235G Supplier TCI America Supplier No. D28235G
Only null left

Chemical Identifiers

CAS 150529-93-4
Molecular Formula C21H22N2O2
Molecular Weight (g/mol) 334.42
MDL Number MFCD00192244
InChI Key JTNVCJCSECAMLD-UHFFFAOYNA-N
Synonym +-2,2'-isopropylidenebis 4r-4-phenyl-2-oxazoline, ph-box, r,r, unii-cg7p67w7w0, r,r-2,2'-isopropylidenebis 4-phenyl-2-oxazoline, r,r-2,2'-dimethylmethylene bis 4-phenyl-2-oxazoline, 4r,4'r-2,2'-propane-2,2-diyl bis 4-phenyl-4,5-dihydrooxazole, oxazole, 2,2'-1-methylethylidene bis 4,5-dihydro-4-phenyl-, 4r,4'r, r,r-2,2-bis 4-phenyl-2-oxazolin-2-yl propane, r,r-2,2-dimethylmethylene bis 4-phenyl-2-oxazoline
PubChem CID 676390
IUPAC Name 4-phenyl-2-[2-(4-phenyl-4,5-dihydro-1,3-oxazol-2-yl)propan-2-yl]-4,5-dihydro-1,3-oxazole
SMILES CC(C)(C1=NC(CO1)C1=CC=CC=C1)C1=NC(CO1)C1=CC=CC=C1

Specifications

Color Yellow
Quantity 5 g
Formula Weight 334.42
Percent Purity ≥96.0% (GC)
Physical Form Liquid
Chemical Name or Material (R,R)-2,2′-Isopropylidenebis(4-phenyl-2-oxazoline)
TSCA No
Recommended Storage Refrigerator