missing translation for 'onlineSavingsMsg'
Learn More

(R)-alpha-Propargylglycine, 98%, 98% ee

Catalog No. AC441221000
Change view
Click to view available options
Quantity:
100 mg
500 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC441221000 100 mg
AC441225000 500 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC441221000 Supplier Thermo Scientific Chemicals Supplier No. 441221000
Only null left

CAS: 23235-03-2 | C5H7NO2 | 113.12 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 23235-03-2
Molecular Formula C5H7NO2
Molecular Weight (g/mol) 113.12
InChI Key DGYHPLMPMRKMPD-SCSAIBSYSA-N
Synonym d-propargylglycine, r-2-aminopent-4-ynoic acid, r-alpha-propargylglycine, unii-2252qef2pl, 4-pentynoic acid, 2-amino-, 2r, 2r-2-aminopent-4-ynoic acid, r-2-propargylglycine, r-2-amino-4-pentynoic acid, d-2-propynylglycine, propargylglycine, d
PubChem CID 6992713
IUPAC Name (2R)-2-aminopent-4-ynoic acid
SMILES C#CCC(C(=O)O)N

Specifications

CAS Min % 97.5
CAS Max % 100.0
Melting Point >180°C
Color White to Yellow
Enantiomeric Excess 98.0% min.
Infrared Spectrum Authentic
Assay Percent Range 97.5 to 102.5%
Packaging Glass bottle
Quantity 100 mg
Specific Rotation 31.65
Formula Weight 113.12
Percent Purity 98%,98% ee
Physical Form Powder
Chemical Name or Material (R)-α-Propargylglycine, 98% ee
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
IF

RUO – Research Use Only