missing translation for 'onlineSavingsMsg'
Learn More

(R)-(-)-Prolinol, 98+%

CAS: 68832-13-3 | C5H11NO | 101.149 g/mol

Supplier:  Thermo Scientific Chemicals L1110906

Quantity 5 g

 View more versions of this product

Catalog No. AAL1110906


Only null left

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

68832-13-3
101.149
HVVNJUAVDAZWCB-RXMQYKEDSA-N
2724541
[(2R)-pyrrolidin-2-yl]methanol
C5H11NO
MFCD00064321
d-prolinol, r-pyrrolidin-2-ylmethanol, 2r-pyrrolidin-2-ylmethanol, r---2-pyrrolidinemethanol, r-prolinol, 2r-pyrrolidin-2-yl methanol, r-1-pyrrolidin-2-yl-methanol, 2-pyrrolidinemethanol, 2r, r---2-hydroxymethyl pyrrolidine
CHEBI:84258
C1CC(NC1)CO

Specifications

(R)-(-)-Prolinol
1.4849
74°C to 76°C (2mmHg)
86°C (186°F)
Air sensitive; Hygroscopic
MFCD00064321
d-prolinol, r-pyrrolidin-2-ylmethanol, 2r-pyrrolidin-2-ylmethanol, r---2-pyrrolidinemethanol, r-prolinol, 2r-pyrrolidin-2-yl methanol, r-1-pyrrolidin-2-yl-methanol, 2-pyrrolidinemethanol, 2r, r---2-hydroxymethyl pyrrolidine
C1CC(NC1)CO
101.149
CHEBI:84258
≥98%
68832-13-3
1.025
5 g
−31° (c=1 in Toluene)
C5H11NO
1523669
HVVNJUAVDAZWCB-RXMQYKEDSA-N
[(2R)-pyrrolidin-2-yl]methanol
2724541
101.15
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H227
Causes skin irritation.
Causes serious eye irritation.
Combustible liquid.

P210-P235-P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P370+P378q-P501c

H227-H315-H319-H335

EINECSNumber : 629-147-4

TSCA : No

Recommended Storage : Ambient temperatures; Store under Nitrogen

SDS
Documents

Documents

Promotions

Promotions