Learn More
(R)-(-)-O-Acetylmandelic acid, 98%
CAS: 51019-43-3 | C10H10O4 | 194.19 g/mol
$55.08 - $166.11
Chemical Identifiers
| CAS | 51019-43-3 |
|---|---|
| Molecular Formula | C10H10O4 |
| Molecular Weight (g/mol) | 194.19 |
| MDL Number | MFCD00004249 |
| InChI Key | OBCUSTCTKLTMBX-UHFFFAOYNA-N |
| Synonym | r-2-acetoxy-2-phenylacetic acid, r---alpha-acetoxyphenylacetic acid, --o-acetyl-d-mandelic acid, r---o-acetylmandelic acid, r-alpha-acetoxyphenylacetic acid, 2r-2-acetyloxy-2-phenylacetic acid, benzeneacetic acid, alpha-acetyloxy-, alphar, r-acetyloxy phenyl acetic acid, s-+-o-acetyl-l-mandelic acid, r---a-acetylmandelic acid |
| PubChem CID | 2733814 |
| IUPAC Name | (2R)-2-acetyloxy-2-phenylacetic acid |
| SMILES | CC(=O)OC(C(O)=O)C1=CC=CC=C1 |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
AAH5649203
|
Thermo Scientific Chemicals
H5649203 |
1 g |
Each for $55.08
|
|
|||||
|
AAH5649206
|
Thermo Scientific Chemicals
H5649206 |
5 g |
Each for $166.11
|
|
|||||
Description
The (R)- and (S)-isomers are chiral derivatizing agents for NMR determination of enantiomeric purity of α-deuterated carboxylic acids, alcohols, and amines.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsThe (R)- and (S)-isomers are chiral derivatizing agents for NMR determination of enantiomeric purity of α-deuterated carboxylic acids, alcohols, and amines.
Solubility
Soluble in methanol. (almost transparency)
Notes
Store in a cool dry place. Incompatible with oxidizing agents. Keep container tightly closed.
Chemical Identifiers
| 51019-43-3 | |
| 194.19 | |
| OBCUSTCTKLTMBX-UHFFFAOYNA-N | |
| 2733814 | |
| CC(=O)OC(C(O)=O)C1=CC=CC=C1 |
| C10H10O4 | |
| MFCD00004249 | |
| r-2-acetoxy-2-phenylacetic acid, r---alpha-acetoxyphenylacetic acid, --o-acetyl-d-mandelic acid, r---o-acetylmandelic acid, r-alpha-acetoxyphenylacetic acid, 2r-2-acetyloxy-2-phenylacetic acid, benzeneacetic acid, alpha-acetyloxy-, alphar, r-acetyloxy phenyl acetic acid, s-+-o-acetyl-l-mandelic acid, r---a-acetylmandelic acid | |
| (2R)-2-acetyloxy-2-phenylacetic acid |
Specifications
| 51019-43-3 | |
| C10H10O4 | |
| 1 g | |
| r-2-acetoxy-2-phenylacetic acid, r---alpha-acetoxyphenylacetic acid, --o-acetyl-d-mandelic acid, r---o-acetylmandelic acid, r-alpha-acetoxyphenylacetic acid, 2r-2-acetyloxy-2-phenylacetic acid, benzeneacetic acid, alpha-acetyloxy-, alphar, r-acetyloxy phenyl acetic acid, s-+-o-acetyl-l-mandelic acid, r---a-acetylmandelic acid | |
| OBCUSTCTKLTMBX-UHFFFAOYNA-N | |
| CC(=O)OC(C(O)=O)C1=CC=CC=C1 | |
| 194.19 | |
| 194.19 | |
| (R)-(-)-O-Acetylmandelic acid |
| 97°C to 99°C | |
| MFCD00004249 | |
| 2694110 | |
| Soluble in methanol. (almost transparency) | |
| −153° (c=2 in Acetone) | |
| (2R)-2-acetyloxy-2-phenylacetic acid | |
| 2733814 | |
| 98% |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P264b-P280i-P305+P351+P338
H319
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only