missing translation for 'onlineSavingsMsg'
Learn More

(R)-(-)-O-Acetylmandelic acid, 98%

Catalog No. AAH5649206
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH5649206 5 g
AAH5649203 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH5649206 Supplier Thermo Scientific Chemicals Supplier No. H5649206
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 51019-43-3 | C10H10O4 | 194.19 g/mol

The (R)- and (S)-isomers are chiral derivatizing agents for NMR determination of enantiomeric purity of α-deuterated carboxylic acids, alcohols, and amines.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
The (R)- and (S)-isomers are chiral derivatizing agents for NMR determination of enantiomeric purity of α-deuterated carboxylic acids, alcohols, and amines.

Solubility
Soluble in methanol. (almost transparency)

Notes
Store in a cool dry place. Incompatible with oxidizing agents. Keep container tightly closed.

Chemical Identifiers

CAS 51019-43-3
Molecular Formula C10H10O4
Molecular Weight (g/mol) 194.19
MDL Number MFCD00004249
InChI Key OBCUSTCTKLTMBX-UHFFFAOYNA-N
Synonym r-2-acetoxy-2-phenylacetic acid, r---alpha-acetoxyphenylacetic acid, --o-acetyl-d-mandelic acid, r---o-acetylmandelic acid, r-alpha-acetoxyphenylacetic acid, 2r-2-acetyloxy-2-phenylacetic acid, benzeneacetic acid, alpha-acetyloxy-, alphar, r-acetyloxy phenyl acetic acid, s-+-o-acetyl-l-mandelic acid, r---a-acetylmandelic acid
PubChem CID 2733814
IUPAC Name (2R)-2-acetyloxy-2-phenylacetic acid
SMILES CC(=O)OC(C(O)=O)C1=CC=CC=C1

Specifications

Melting Point 97°C to 99°C
Quantity 5 g
Beilstein 2694110
Solubility Information Soluble in methanol. (almost transparency)
Optical Rotation −153° (c=2 in Acetone)
Formula Weight 194.19
Percent Purity 98%
Chemical Name or Material (R)-(-)-O-Acetylmandelic acid
Hazard Category H319
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P264b-P280i-P305+P351+P338
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only