missing translation for 'onlineSavingsMsg'
Learn More

(R)-(+)-NBD-Pro-COCl [=(R)-(+)-4-Nitro-7-(2-chloroformylpyrrolidin-1-yl)-2,1,3-benzoxadiazole], TCI America™

Catalog No. A5566100MG
Change view
Click to view available options
Quantity:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A5566100MG 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. A5566100MG Supplier TCI America Supplier No. A5566100MG

[HPLC Labeling Reagent for e.e.Determination]

Chemical Identifiers

CAS 159717-69-8
Molecular Formula C11H9ClN4O4
Molecular Weight (g/mol) 296.667
MDL Number MFCD00191511
InChI Key XBQYXPYABMCADZ-MRVPVSSYSA-N
Synonym (R)-(+)-4-Nitro-7-(2-chloroformylpyrrolidin-1-yl)benzofurazan, (R)-(+)-4-Nitro-7-(2-chloroformylpyrrolidin-1-yl)-2,1,3-benzoxadiazole
PubChem CID 44630075
IUPAC Name (2R)-1-(4-nitro-2,1,3-benzoxadiazol-7-yl)pyrrolidine-2-carbonyl chloride
SMILES C1CC(N(C1)C2=CC=C(C3=NON=C23)[N+](=O)[O-])C(=O)Cl

Specifications

Quantity 100 mg
Formula Weight 296.67
Physical Form Crystalline Powder
Chemical Name or Material (R)-(+)-NBD-Pro-COCl [=(R)-(+)-4-Nitro-7-(2-chloroformylpyrrolidin-1-yl)-2,1,3-benz
TSCA No
Recommended Storage Refrigerator