missing translation for 'onlineSavingsMsg'
Learn More

(R)-(+)-N,N-Dimethyl-1-phenylethylamine 98.0+%, TCI America™

Catalog No. D17071ML
Change view
Click to view available options
Quantity:
1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D17071ML 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. D17071ML Supplier TCI America Supplier No. D17071ML

Chemical Identifiers

CAS 19342-01-9
Molecular Formula C10H15N
Molecular Weight (g/mol) 149.237
MDL Number MFCD00008857
InChI Key BVURNMLGDQYNAF-SECBINFHSA-N
Synonym r-+-n,n-dimethyl-1-phenylethylamine, r-n,n-dimethyl-1-phenylethanamine, unii-13x1wu55oq, 1r-n,n-dimethyl-1-phenylethanamine, r-+-n,n-dimethyl-1-phenylethanamine, dimethyl 1r-1-phenylethyl amine, n,n-dimethyl-1-phenylethanamine #, r-+-n,n-dimethyl-1-phenethylamine
PubChem CID 103034
IUPAC Name (1R)-N,N-dimethyl-1-phenylethanamine
SMILES CC(C1=CC=CC=C1)N(C)C

Specifications

Color Yellow
Boiling Point 94°C
Quantity 1 mL
Formula Weight 149.24
Percent Purity ≥98.0% (T)
Physical Form Liquid
Chemical Name or Material (R)-(+)-N,N-Dimethyl-1-phenylethylamine
EINECSNumber (3)-0318
TSCA No