missing translation for 'onlineSavingsMsg'
Learn More

(R)-N-Glycidylphthalimide 99.0+%, TCI America™

Catalog No. G03275G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
G03275G 5 g
G032725G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. G03275G Supplier TCI America Supplier No. G03275G
Only null left

Chemical Identifiers

CAS 181140-34-1
Molecular Formula C11H9NO3
Molecular Weight (g/mol) 203.197
MDL Number MFCD04973349
InChI Key DUILGEYLVHGSEE-SSDOTTSWSA-N
Synonym (R)-N-(2,3-Epoxypropyl)phthalimide
PubChem CID 719415
IUPAC Name 2-[[(2R)-oxiran-2-yl]methyl]isoindole-1,3-dione
SMILES C1C(O1)CN2C(=O)C3=CC=CC=C3C2=O

Specifications

Melting Point 101°C
Color White
Quantity 5 g
Formula Weight 203.20
Percent Purity ≥99.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material (R)-N-Glycidylphthalimide