missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Chemical Identifiers
| CAS | 220497-85-8 |
|---|---|
| Molecular Formula | C22H19NO5 |
| Molecular Weight (g/mol) | 377.396 |
| MDL Number | MFCD01311774 |
| InChI Key | AJXDCHXGNUFBRC-HXUWFJFHSA-N |
| Synonym | fmoc-d-2-furylalanine, r-2-9h-fluoren-9-yl methoxy carbonyl amino-3-furan-2-yl propanoic acid, fmoc-beta-2-furyl-d-ala-oh, n-9h-fluoren-9-ylmethoxy carbonyl-3-2-furyl-d-alanine, 2r-2-9h-fluoren-9-ylmethoxy carbonyl amino-3-furan-2-yl propanoic acid, fmoc-3-2-furyl-d-alanine, n-fmoc-3-2-furyl-d-alanine, fmoc-beta-2-furyl-d-ala-oh hplc |
| PubChem CID | 16211163 |
| IUPAC Name | (2R)-2-(9H-fluoren-9-ylmethoxycarbonylamino)-3-(furan-2-yl)propanoic acid |
| SMILES | C1=CC=C2C(=C1)C(C3=CC=CC=C32)COC(=O)NC(CC4=CC=CO4)C(=O)O |
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity & Availability | ||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity & Availability | ||||
|
AC362495000
|
N/A | 500mg | Glass bottle |
N/A
|
N/A | ||||
| Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
Chemical Identifiers
| 220497-85-8 | |
| 377.396 | |
| AJXDCHXGNUFBRC-HXUWFJFHSA-N | |
| 16211163 | |
| C1=CC=C2C(=C1)C(C3=CC=CC=C32)COC(=O)NC(CC4=CC=CO4)C(=O)O |
| C22H19NO5 | |
| MFCD01311774 | |
| fmoc-d-2-furylalanine, r-2-9h-fluoren-9-yl methoxy carbonyl amino-3-furan-2-yl propanoic acid, fmoc-beta-2-furyl-d-ala-oh, n-9h-fluoren-9-ylmethoxy carbonyl-3-2-furyl-d-alanine, 2r-2-9h-fluoren-9-ylmethoxy carbonyl amino-3-furan-2-yl propanoic acid, fmoc-3-2-furyl-d-alanine, n-fmoc-3-2-furyl-d-alanine, fmoc-beta-2-furyl-d-ala-oh hplc | |
| (2R)-2-(9H-fluoren-9-ylmethoxycarbonylamino)-3-(furan-2-yl)propanoic acid |
Specifications
| 220497-85-8 | |
| 100.0 | |
| Authentic | |
| C22H19NO5 | |
| fmoc-d-2-furylalanine, r-2-9h-fluoren-9-yl methoxy carbonyl amino-3-furan-2-yl propanoic acid, fmoc-beta-2-furyl-d-ala-oh, n-9h-fluoren-9-ylmethoxy carbonyl-3-2-furyl-d-alanine, 2r-2-9h-fluoren-9-ylmethoxy carbonyl amino-3-furan-2-yl propanoic acid, fmoc-3-2-furyl-d-alanine, n-fmoc-3-2-furyl-d-alanine, fmoc-beta-2-furyl-d-ala-oh hplc | |
| C1=CC=C2C(=C1)C(C3=CC=CC=C32)COC(=O)NC(CC4=CC=CO4)C(=O)O | |
| 377.396 | |
| 377.39 | |
| (R)-N-FMOC-(2-Furyl)alanine, 95%, (98% e.e.) |
| 94.0 | |
| 97.5% min. (HPLC) | |
| 95% | |
| MFCD01311774 | |
| AJXDCHXGNUFBRC-HXUWFJFHSA-N | |
| (2R)-2-(9H-fluoren-9-ylmethoxycarbonylamino)-3-(furan-2-yl)propanoic acid | |
| 16211163 | |
| ≥94% |
Safety and Handling
TSCA : TSCA
RUO – Research Use Only