missing translation for 'onlineSavingsMsg'
Learn More

(R)-(+)-N-BOC-2-Piperidinecarboxylic acid, 98%, Thermo Scientific™

Supplier:  Thermo Scientific Chemicals 399250050

Quantity 5g
Packaging Glass bottle

 View more versions of this product

Catalog No. AC399250050

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description

Description

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications

Chemical Identifiers

28697-17-8
229.27
JQAOHGMPAAWWQO-MRVPVSSYSA-N
688618
CC(C)(C)OC(=O)N1CCCCC1C(=O)O
C11H19NO4
MFCD00237380
r-+-n-boc-2-piperidinecarboxylic acid, boc-d-hopro-oh, boc-d-pipecolic acid, r-1-tert-butoxycarbonyl piperidine-2-carboxylic acid, boc-d-pip-oh, d-n-boc-pipecolic acid, r-1-n-boc-pipecolinic acid, r-n-boc-piperidine-2-carboxylic acid, +-n-boc-r-pipecolinic acid, r-n-boc-2-piperidinecarboxylic acid
(2R)-1-[(2-methylpropan-2-yl)oxycarbonyl]piperidine-2-carboxylic acid

Specifications

(R)-(+)-N-BOC-2-Piperidinecarboxylic acid
97.5
Authentic
Glass bottle
5g
r-+-n-boc-2-piperidinecarboxylic acid, boc-d-hopro-oh, boc-d-pipecolic acid, r-1-tert-butoxycarbonyl piperidine-2-carboxylic acid, boc-d-pip-oh, d-n-boc-pipecolic acid, r-1-n-boc-pipecolinic acid, r-n-boc-piperidine-2-carboxylic acid, +-n-boc-r-pipecolinic acid, r-n-boc-2-piperidinecarboxylic acid
JQAOHGMPAAWWQO-MRVPVSSYSA-N
(2R)-1-[(2-methylpropan-2-yl)oxycarbonyl]piperidine-2-carboxylic acid
688618
98%
28697-17-8
100.0
98%
C11H19NO4
MFCD00237380
+62.0° to +65.0° (20°C, 589nm) (c=1%, acetic acid)
CC(C)(C)OC(=O)N1CCCCC1C(=O)O
229.27
229.27
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement:
May cause respiratory irritation.
Harmful if swallowed.
Causes serious eye irritation.
Causes skin irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

SDS
Documents

Documents

Promotions

Promotions

Promo
Expires: 06-30-2027
Get Up to $1,800 in Chemicals with Purchase
Spend $250 or more on Thermo Fisher Scientific chemicals and get up to $1,800 to spend on additional chemicals.

Adding...