missing translation for 'onlineSavingsMsg'
Learn More

(R)-N-Boc-2-hydroxymethylmorpholine, 99%

Catalog No. AAH6006203
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH6006203 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAH6006203 Supplier Thermo Scientific Chemicals Supplier No. H6006203
Only null left

CAS: 135065-71-3 | C10H19NO4 | 217.27 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 135065-71-3
Molecular Formula C10H19NO4
Molecular Weight (g/mol) 217.27
MDL Number MFCD09260605
InChI Key FJYBLMJHXRWDAQ-UHFFFAOYNA-N
Synonym r-n-boc-2-hydroxymethylmorpholine, r-tert-butyl 2-hydroxymethyl morpholine-4-carboxylate, tert-butyl 2r-2-hydroxymethyl morpholine-4-carboxylate, r-4-boc-2-hydroxymethyl-morpholine, r-2-hydroxymethylmorpholine-4-carboxylic acid tert-butyl ester, n-boc-r-morpholin-2-yl methanol, r-2-hydroxymethyl-morpholine-4-carboxylic acid tert-butyl ester, ksc496k2p, r-4-n-boc-2-morpholinemethanol, r-n-boc-2-hydroxymethyl morpholine
PubChem CID 1512576
IUPAC Name tert-butyl (2R)-2-(hydroxymethyl)morpholine-4-carboxylate
SMILES CC(C)(C)OC(=O)N1CCOC(CO)C1

Specifications

Quantity 1 g
Formula Weight 217.27
Percent Purity 99%
Chemical Name or Material (R)-N-Boc-2-hydroxymethylmorpholine
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only