missing translation for 'onlineSavingsMsg'
Learn More

(R)-N-Boc-1,2,3,4-tetrahydroisoquinoline-3-carboxylic acid, 98%

Catalog No. p-7034294
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH5196503 1 g
AAH5196506 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH5196503 Supplier Thermo Scientific Chemicals Supplier No. H5196503
Only null left

CAS: 115962-35-1 | C15H18NO4 | 276.31 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 115962-35-1
Molecular Formula C15H18NO4
Molecular Weight (g/mol) 276.31
MDL Number MFCD00143818
InChI Key HFPVZPNLMJDJFB-GFCCVEGCSA-M
Synonym boc-d-tic-oh, r-2-tert-butoxycarbonyl-1,2,3,4-tetrahydroisoquinoline-3-carboxylic acid, boc-d-1,2,3,4-tetrahydroisoquinoline-3-carboxylic acid, n-boc-d-1,2,3,4-tetrahydroisoquinoline-3-carboxylic acid, 3r-2-tert-butoxy carbonyl-1,2,3,4-tetrahydroisoquinoline-3-carboxylic acid, r---n-boc-1,2,3,4-tetrahydro-3-isoquinolinecarboxylic acid, 3r-2-tert-butoxycarbonyl-3,4-dihydro-1h-isoquinoline-3-carboxylic acid, r---2-tertbutoxycarbonyl-1,2,3,4-tetrahydro-3-isoquinolinecarboxylic acid, boc-d-tic, ambotzbaa1055
PubChem CID 688607
IUPAC Name (3R)-2-[(2-methylpropan-2-yl)oxycarbonyl]-3,4-dihydro-1H-isoquinoline-3-carboxylic acid
SMILES CC(C)(C)OC(=O)N1CC2=CC=CC=C2C[C@@H]1C([O-])=O

Specifications

Quantity 1 g
Formula Weight 277.32
Percent Purity 98%
Chemical Name or Material (R)-N-Boc-1,2,3,4-tetrahydroisoquinoline-3-carboxylic acid
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only