missing translation for 'onlineSavingsMsg'
Learn More

(R)-(-)-N-(3,5-Dinitrobenzoyl)-alpha-phenylethylamine, TCI America™

Catalog No. D18521G
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D18521G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. D18521G Supplier TCI America Supplier No. D18521G
Only null left

Chemical Identifiers

CAS 69632-32-2
Molecular Formula C15H13N3O5
Molecular Weight (g/mol) 315.285
MDL Number MFCD00010866
InChI Key ABEVDCGKLRIYRW-SNVBAGLBSA-N
Synonym r---n-3,5-dinitrobenzoyl-alpha-phenylethylamine, 3,5-dinitro-n-1r-1-phenylethyl benzamide, r---3,5-dinitro-n-1-phenylethyl benzamide, 3,5-dinitro-n-1-phenylethyl benzamide #, n-r-1-phenylethyl-3,5-dinitrobenzamide, r---n-3,5-dinitrobenzoyl-1-phenylethylamine, r---n-3,5-dinitrobenzoyl-.alpha.-methylbenzylamine, 3-hydroxyazonoyl-5-1r-1-phenylethyl carbamoyl phenyl azinic acid
PubChem CID 2794133
IUPAC Name 3,5-dinitro-N-[(1R)-1-phenylethyl]benzamide
SMILES CC(C1=CC=CC=C1)NC(=O)C2=CC(=CC(=C2)[N+](=O)[O-])[N+](=O)[O-]

Specifications

Melting Point 159°C
Color White-Yellow
Quantity 1 g
Formula Weight 315.29
Physical Form Crystalline Powder
Chemical Name or Material (R)-(-)-N-(3,5-Dinitrobenzoyl)-alpha-phenylethylamine