missing translation for 'onlineSavingsMsg'
Learn More

(R)-(+)-α-Methylbenzyl isocyanate, ≥99.0% (sum of enantiomers, GC), MilliporeSigma™ Supelco™

Catalog No. 111018734
Change view
Click to view available options
Quantity:
1 mL
5 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111018734 1 mL
111018022 5 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 111018734 Supplier Merck Emd Millipore Supplier No. 779681ML

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

(R)-(+)-a-Methylbenzyl isocyanate [(R)-(+)-MBIC] is a derivatizing agent.

Chemical Identifiers

MDL Number MFCD00063015
Synonym (R)-(+)-1-Phenylethyl isocyanate

Specifications

CAS 33375-06-3
Density 1.045 g/mL at 20°C; 1.046 g/mL (at 20°C)
Boiling Point 55°C to 56°C/2.5 mmHg
Flash Point 65°C - Closed Cup
Packaging Glass Bottle
Linear Formula C6H5 CH(CH3)NCO
Quantity 1 mL
Formula Weight 147.17
Percent Purity ≥99.0% (sum of enantiomers, GC); ≥99.0%
Grade For chiral derivatization