missing translation for 'onlineSavingsMsg'
Learn More

(R)-(-)-Mandelic acid, 99%

Catalog No. p-154642
Change view
missing translation for 'orderingAttributeHoverText'
Quantity:
100 g
25 g
Packaging:
Plastic bottle
Catalog No. Quantity Packaging
AC125320250 25 g Plastic bottle
AC125321000 100 g Plastic bottle
2 options

Catalog No. AC125320250

Supplier: Thermo Scientific Chemicals 125320250

Only null left

CAS: 611-71-2 | C8H8O3 | 152.15 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 611-71-2
Molecular Formula C8H8O3
Molecular Weight (g/mol) 152.15
MDL Number MFCD00064251
InChI Key IWYDHOAUDWTVEP-SSDOTTSWSA-N
Synonym r-mandelic acid, r---mandelic acid, d-mandelic acid, r-2-hydroxy-2-phenylacetic acid, --mandelic acid, d--mandelic acid, d---mandelic acid, 2r-2-hydroxy-2-phenylacetic acid, r-alpha-hydroxyphenylacetic acid, d-2-phenylglycolic acid
PubChem CID 11914
ChEBI CHEBI:17656
IUPAC Name (2R)-2-hydroxy-2-phenylacetic acid
SMILES O[C@@H](C(O)=O)C1=CC=CC=C1

Specifications

Melting Point 130°C to 135°C
CAS Min % 98.5
Color Beige-Yellow to White
CAS Max % 100.0
Assay Percent Range 99%
Linear Formula C6H5CH(OH)CO2H
Formula Weight 152.15
Percent Purity ≥98.5%
Quantity 25 g
Infrared Spectrum Authentic
Packaging Plastic bottle
Physical Form Crystalline Powder, Crystals or Flakes
Chemical Name or Material (R)-(-)-Mandelic acid, 99%
Show More Show Less
Hazard Category LIGHT SENSITIVE
Hazard Statement GHS H Statement:
Causes serious eye damage.
Precautionary Statement GHS P Statement:
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber 210-276-6
TSCA TSCA

RUO – Research Use Only