missing translation for 'onlineSavingsMsg'
Learn More

(R)-(+)-Indoline-2-carboxylic acid, 97%, Thermo Scientific Chemicals

$181.82 - $181.82

Chemical Identifiers

CAS 98167-06-7
Molecular Formula C9H9NO2
Molecular Weight (g/mol) 163.176
MDL Number MFCD00792496
InChI Key QNRXNRGSOJZINA-MRVPVSSYSA-N
Synonym r-indoline-2-carboxylic acid, r-+-indoline-2-carboxylic acid, 2r-2,3-dihydro-1h-indole-2-carboxylic acid, r---indoline-2-carboxylic acid, 2r-indoline-2-carboxylic acid, r-+-2,3-dihydroindole-2-carboxylic acid, r-2,3-dihydro-1h-indole-2-carboxylic acid, 1h-indole-2-carboxylic acid, 2,3-dihydro-, 2r, pubchem20604
PubChem CID 6928266
IUPAC Name (2R)-2,3-dihydro-1H-indole-2-carboxylic acid
SMILES C1C(NC2=CC=CC=C21)C(=O)O
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAH2762603
View Documents
Thermo Scientific Chemicals
H2762603
1 g N/A
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

98167-06-7
163.176
QNRXNRGSOJZINA-MRVPVSSYSA-N
6928266
C1C(NC2=CC=CC=C21)C(=O)O
C9H9NO2
MFCD00792496
r-indoline-2-carboxylic acid, r-+-indoline-2-carboxylic acid, 2r-2,3-dihydro-1h-indole-2-carboxylic acid, r---indoline-2-carboxylic acid, 2r-indoline-2-carboxylic acid, r-+-2,3-dihydroindole-2-carboxylic acid, r-2,3-dihydro-1h-indole-2-carboxylic acid, 1h-indole-2-carboxylic acid, 2,3-dihydro-, 2r, pubchem20604
(2R)-2,3-dihydro-1H-indole-2-carboxylic acid

Specifications

98167-06-7
C9H9NO2
1 g
QNRXNRGSOJZINA-MRVPVSSYSA-N
(2R)-2,3-dihydro-1H-indole-2-carboxylic acid
6928266
97%
∽177°C (decomposition)
MFCD00792496
r-indoline-2-carboxylic acid, r-+-indoline-2-carboxylic acid, 2r-2,3-dihydro-1h-indole-2-carboxylic acid, r---indoline-2-carboxylic acid, 2r-indoline-2-carboxylic acid, r-+-2,3-dihydroindole-2-carboxylic acid, r-2,3-dihydro-1h-indole-2-carboxylic acid, 1h-indole-2-carboxylic acid, 2,3-dihydro-, 2r, pubchem20604
C1C(NC2=CC=CC=C21)C(=O)O
163.176
163.18
(R)-(+)-Indoline-2-carboxylic acid
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H361-H373-H317
Suspected of damaging fertility or the unborn child.
May cause damage to organs through prolonged or repeated exposure.
May cause an allergic skin reaction.

P201-P202-P260-P272-P280g-P281-P302+P352-P308+P313-P333+P313-P363-P501c

H317-H361f-H373

TSCA : No

Recommended Storage : Ambient temperatures

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only