missing translation for 'onlineSavingsMsg'
Learn More

(R)-(+)-Indoline-2-carboxylic acid, 97%, Thermo Scientific Chemicals

Catalog No. AAH2762603
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH2762603 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAH2762603 Supplier Thermo Scientific Chemicals Supplier No. H2762603
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 98167-06-7 | C9H9NO2 | 163.176 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 98167-06-7
Molecular Formula C9H9NO2
Molecular Weight (g/mol) 163.176
MDL Number MFCD00792496
InChI Key QNRXNRGSOJZINA-MRVPVSSYSA-N
Synonym r-indoline-2-carboxylic acid, r-+-indoline-2-carboxylic acid, 2r-2,3-dihydro-1h-indole-2-carboxylic acid, r---indoline-2-carboxylic acid, 2r-indoline-2-carboxylic acid, r-+-2,3-dihydroindole-2-carboxylic acid, r-2,3-dihydro-1h-indole-2-carboxylic acid, 1h-indole-2-carboxylic acid, 2,3-dihydro-, 2r, pubchem20604
PubChem CID 6928266
IUPAC Name (2R)-2,3-dihydro-1H-indole-2-carboxylic acid
SMILES C1C(NC2=CC=CC=C21)C(=O)O

Specifications

Melting Point ∽177°C (decomposition)
Quantity 1 g
Formula Weight 163.18
Percent Purity 97%
Chemical Name or Material (R)-(+)-Indoline-2-carboxylic acid
Hazard Category H317-H361f-H373
Hazard Statement GHS H Statement
H361-H373-H317
Suspected of damaging fertility or the unborn child.
May cause damage to organs through prolonged or repeated exposure.
May cause an allergic skin reaction.
Precautionary Statement P201-P202-P260-P272-P280g-P281-P302+P352-P308+P313-P333+P313-P363-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only