missing translation for 'onlineSavingsMsg'
Learn More

(R)-(-)-Glycidyl nosylate, 98%

Catalog No. ac296410025
Change view
Click to view available options
Quantity:
2.5 g
10 g
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC296410025 2.5 g Glass bottle
AC296410100 10 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC296410025 Supplier Thermo Scientific Chemicals Supplier No. 296410025
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 115314-17-5 | C9H9NO6S | 259.23 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 115314-17-5
Molecular Formula C9H9NO6S
Molecular Weight (g/mol) 259.23
MDL Number MFCD00013436
InChI Key AIHIHVZYAAMDPM-MRVPVSSYSA-N
Synonym r---glycidyl nosylate, r-glycidyl nosylate, r-oxiran-2-ylmethyl 3-nitrobenzenesulfonate, r-glycidyl 3-nitrobenzenesulfonate, r---glycidyl 3-nitrobenzenesulfonate, r---glycidyl-3-nitrobenzenesulfonate, ccris 6393, oxiranylmethyl r-3-nitrobenzenesulfonate, benzenesulfonic acid, 3-nitro-, oxiranylmethyl ester, r
PubChem CID 146513
IUPAC Name [(2R)-oxiran-2-yl]methyl 3-nitrobenzenesulfonate
SMILES C1C(O1)COS(=O)(=O)C2=CC=CC(=C2)[N+](=O)[O-]

Specifications

CAS Min % 97.5
CAS Max % 100.0
Melting Point 59°C to 63°C
Color Yellow
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Quantity 2.5 g
Specific Rotation Condition −21.5° (20°C c=2,CHCl3)
Specific Rotation -21.5
Formula Weight 259.23
Percent Purity 98%
Physical Form Crystalline Needles or Powder
Chemical Name or Material (R)-(-)-Glycidyl nosylate, 98%
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
RTECSNumber DB7192000
TSCA TSCA

RUO – Research Use Only