missing translation for 'onlineSavingsMsg'
Learn More

(R)-(-)-Epichlorohydrin, 98+%

Catalog No. AAL1419803
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL1419803 1 g
AAL1419806 5 g
AAL1419814 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAL1419803 Supplier Thermo Scientific Chemicals Supplier No. L1419803
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 51594-55-9 | C3H5ClO | 92.52 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 51594-55-9
Molecular Formula C3H5ClO
Molecular Weight (g/mol) 92.52
MDL Number MFCD00077759
InChI Key BRLQWZUYTZBJKN-VKHMYHEASA-N
Synonym r---epichlorohydrin, r-epichlorohydrin, 2r-2-chloromethyl oxirane, --epichlorohydrin, r-chloromethyl oxirane, r-2-chloromethyl oxirane, epichlorohydrin,-, unii-vec01h609i, r-1-chloro-2,3-epoxypropane
PubChem CID 2734062
ChEBI CHEBI:18662
IUPAC Name (2R)-2-(chloromethyl)oxirane
SMILES ClC[C@H]1CO1

Specifications

Density 1.18
Boiling Point 116°C to 117°C
Flash Point 32°C (90°F)
Refractive Index 1.438
Quantity 1 g
UN Number UN2023
Beilstein 1420785
Optical Rotation −35° (c=1 in Methanol)
Formula Weight 92.53
Percent Purity ≥98%
Chemical Name or Material (R)-(-)-Epichlorohydrin
Show More Show Less
missing translation for 'healthHazard1' H226-H301+H311-H314-H317-H350
missing translation for 'healthHazard2' GHS H Statement
H301-H311-H331-H350-H314-H226-H317
Toxic if swallowed.
Toxic in contact with skin.
Toxic if inhaled.
May cause cancer.
Causes severe skin burns and eye damage.
Flammable liquid and vapor.
May cause an allergic skin reaction.
missing translation for 'healthHazard3' P201-P202-P210-P233-P240-P241-P242-P243-P260-P264b-P270-P272-P280g-P281-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P333+P313-P363-P370+P378q-P501c
DOTInformation Transport Hazard Class: 6.1; Packing Group: II; Proper Shipping Name: EPICHLOROHYDRIN
EINECSNumber 424-280-2
RTECSNumber RR0427000
TSCA Yes
Recommended Storage Keep cold

RUO – Research Use Only