missing translation for 'onlineSavingsMsg'
Learn More

(R)-(-)-Deprenyl hydrochloride

Catalog No. p-7042078
Change view
Click to view available options
Quantity:
250 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAJ61286MD 250 mg
AAJ6128603 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAJ61286MD Supplier Thermo Scientific Chemicals Supplier No. J61286MD
Only null left

Selective MAO-B inhibitor; anti-Parkinsonian agent | CAS: 14611-52-0 | C13H18ClN | 223.74 g/mol

(R)-(-)-Deprenyl hydrochloride provides neuroprotection which may be relevant to Parkinson’s disease, Alzheimer’s disease, Huntington’s disease, and stroke. It is an inhibitor of MAO-B (monoamine oxidase-B) which demonstrates extensive neuroprotective effects shown to be uncorrelated with MAO-B inhibiting activity. It demonstrates protection of dopaminergic neurons from excitotoxicity mediated by NMDA activation. It also demonstrates decreases in apoptosis of PC12 neuronal cells accompanied with an induction of new protein synthesis through direct transcriptional and translational interaction, and reduced cell death and internucleosomal DNA degredation at concentrations too low to inhibit MAO-B.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
(R)-(-)-Deprenyl hydrochloride provides neuroprotection which may be relevant to Parkinson’s disease, Alzheimer’s disease, Huntington’s disease, and stroke. It is an inhibitor of MAO-B (monoamine oxidase-B) which demonstrates extensive neuroprotective effects shown to be uncorrelated with MAO-B inhibiting activity. It demonstrates protection of dopaminergic neurons from excitotoxicity mediated by NMDA activation. It also demonstrates decreases in apoptosis of PC12 neuronal cells accompanied with an induction of new protein synthesis through direct transcriptional and translational interaction, and reduced cell death and internucleosomal DNA degredation at concentrations too low to inhibit MAO-B.

Solubility
Soluble in water at 100mM

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.

Chemical Identifiers

CAS 14611-52-0
Molecular Formula C13H18ClN
Molecular Weight (g/mol) 223.74
MDL Number MFCD00069299
InChI Key IYETZZCWLLUHIJ-UTONKHPSSA-N
Synonym selegiline hydrochloride, eldepryl, zelapar, selegiline hcl, l-deprenyl hydrochloride, jumex hydrochloride, --deprenyl hydrochloride, eldepryl hydrochloride, r-n-methyl-n-1-phenylpropan-2-yl prop-2-yn-1-amine hydrochloride, zydis selegiline
PubChem CID 26758
ChEBI CHEBI:9087
IUPAC Name hydrogen methyl[(2R)-1-phenylpropan-2-yl](prop-2-yn-1-yl)amine chloride
SMILES [H+].[Cl-].C[C@H](CC1=CC=CC=C1)N(C)CC#C

Specifications

Color White
Quantity 250 mg
Merck Index 14,8428
Solubility Information Soluble in water at 100mM
Optical Rotation −12° (c=1 in Water)
Formula Weight 223.74
Physical Form Crystalline Powder
Chemical Name or Material (R)-(-)-Deprenyl hydrochloride
Hazard Category H315-H319
Hazard Statement GHS H Statement
H315-H319
Causes skin irritation.
Causes serious eye irritation.
Precautionary Statement P264b-P280-P302+P352-P305+P351+P338-P332+P313-P362
RTECSNumber DA0292500
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only