missing translation for 'onlineSavingsMsg'
Learn More

(R)-(-)-Carvone 99.0+%, TCI America™

Catalog No. C070325ML
Change view
Click to view available options
Quantity:
25 mL
100 mL
500 mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C070325ML 25 mL
C0703100ML 100 mL
C0703500ML 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. C070325ML Supplier TCI America Supplier No. C070325ML

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 6485-40-1
Molecular Formula C10H14O
Molecular Weight (g/mol) 150.22
MDL Number MFCD00001578
InChI Key ULDHMXUKGWMISQ-SECBINFHSA-N
Synonym --carvone, l-carvone, r---carvone, l--carvone, r-2-methyl-5-prop-1-en-2-yl cyclohex-2-enone, 4r-carvone, levo-carvone, --p-mentha-6,8-dien-2-one, --r-carvone
PubChem CID 439570
ChEBI CHEBI:15400
IUPAC Name (5R)-2-methyl-5-(prop-1-en-2-yl)cyclohex-2-en-1-one
SMILES CC(=C)[C@@H]1CC=C(C)C(=O)C1

Specifications

Color Yellow
Boiling Point 231°C
Quantity 25 mL
Formula Weight 150.22
Percent Purity ≥99.0% (GC)
Physical Form Liquid
Chemical Name or Material (R)-(-)-Carvone
EINECSNumber (3)-2402
RTECSNumber OS8650000
TSCA Yes