missing translation for 'onlineSavingsMsg'
Learn More

(R)-(-)-Carvone, 98%

CAS: 6485-40-1 | C10H14O | 150.22 g/mol

Supplier:  Thermo Scientific Chemicals A139000B

Quantity 1000 g

 View more versions of this product

Catalog No. AAA139000B


Only null left

Description

Description

(R)-(-)-Carvone is used in the flavor and food industry such as chewing gum additives. It is used in air freshening products such as in essential oils as well as in aromatherapy and alternative medicine. It is used to prepare carvomenthol, carvomenthone, dihydrocarvone, carveol and limonene. It reacts with lithium dimethylcuprate to place a methyl group trans to the isopropenyl group with good stereoselectivity. It is also used as a mosquito repellent and prevent premature sprouting of potatoes. Further, it is employed as an important starting material for the synthesis of enantiopure (R)-(+)-3-methyl-6-isopropenyl-cyclohept-3-enone-1 and (4S,6R,7R)-trihydroxy-1-octyne derivatives. It is utilized as a vital raw material for the asymmetric total synthesis of natural products. In addition to this, it is used as a chiral starting material.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
(R)-(-)-Carvone is used in the flavor and food industry such as chewing gum additives. It is used in air freshening products such as in essential oils as well as in aromatherapy and alternative medicine. It is used to prepare carvomenthol, carvomenthone, dihydrocarvone, carveol and limonene. It reacts with lithium dimethylcuprate to place a methyl group trans to the isopropenyl group with good stereoselectivity. It is also used as a mosquito repellent and prevent premature sprouting of potatoes. Further, it is employed as an important starting material for the synthesis of enantiopure (R)-(+)-3-methyl-6-isopropenyl-cyclohept-3-enone-1 and (4S,6R,7R)-trihydroxy-1-octyne derivatives. It is utilized as a vital raw material for the asymmetric total synthesis of natural products. In addition to this, it is used as a chiral starting material.

Solubility
Miscible with alcohol, ether, chloroform, propylene glycol and mineral oils. Immiscible with water and glycerol.

Notes
Store in a cool place. Incompatible with strong oxidizing agents.
Specifications

Chemical Identifiers

6485-40-1
150.22
ULDHMXUKGWMISQ-SECBINFHSA-N
439570
CC(=C)[C@@H]1CC=C(C)C(=O)C1
C10H14O
MFCD00001578
--carvone, l-carvone, r---carvone, l--carvone, r-2-methyl-5-prop-1-en-2-yl cyclohex-2-enone, 4r-carvone, levo-carvone, --p-mentha-6,8-dien-2-one, --r-carvone
CHEBI:15400

Specifications

(R)-(-)-Carvone
0.959
93°C (199°F)
98%
1.499
MFCD00001578
--carvone, l-carvone, r---carvone, l--carvone, r-2-methyl-5-prop-1-en-2-yl cyclohex-2-enone, 4r-carvone, levo-carvone, --p-mentha-6,8-dien-2-one, --r-carvone
ULDHMXUKGWMISQ-SECBINFHSA-N
−61° (Neat)
150.22
CHEBI:15400
98%
6485-40-1
228°C to 230°C
Pleasant
C10H14O
1000 g
2206714
Miscible with alcohol,ether,chloroform,propylene glycol and mineral oils. Immiscible with water and glycerol.
CC(=C)[C@@H]1CC=C(C)C(=O)C1
(5R)-2-methyl-5-(prop-1-en-2-yl)cyclohex-2-en-1-one
439570
150.22
Liquid
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
H302-H227
Harmful if swallowed.
Combustible liquid.

P210-P261-P272-P280-P302+P352-P333+P313-P363-P370+P378q-P501c

H227-H317

EINECSNumber : 229-352-5

RTECSNumber : OS8650000

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions