missing translation for 'onlineSavingsMsg'
Learn More

(R)-(+)-Butylene Oxide 98.0+%, TCI America™

Catalog No. p-7123190
Change view
Click to view available options
Quantity:
5 mL
25 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B38325ML 5 mL
B383225ML 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B38325ML Supplier TCI America Supplier No. B38325ML

Chemical Identifiers

CAS 3760-95-0
Molecular Formula C4H8O
Molecular Weight (g/mol) 72.107
MDL Number MFCD02683444
InChI Key RBACIKXCRWGCBB-SCSAIBSYSA-N
Synonym r-+-1,2-epoxybutane, r-2-ethyloxirane, 2r-2-ethyloxirane, r-1,2-epoxybutane, oxirane, ethyl-, 2r, r-epoxybutane, pubchem14098, r-+-ethyloxirane, r-+-butylene oxide
PubChem CID 11147670
IUPAC Name (2R)-2-ethyloxirane
SMILES CCC1CO1

Specifications

Color Colorless
Boiling Point 63°C
Quantity 5 mL
UN Number 3022
Formula Weight 72.11
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material (R)-(+)-Butylene Oxide
EINECSNumber (2)-0229
TSCA No