Learn More
Electron Microscopy Sciences R-Buffer U (10x), 250 ml for use in Retriever Thermal Slide Processor 250 ML
Catalog No. 5031180
Encompass
Used with the Retriever benchtop thermal slide processer that is used on formalin-fixed, paraffin embedded tissues prior to immunostaining.
Catalog No. 5031180 Supplier Electron Microscopy Sciences Supplier No. 6270613
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Used with the Retriever benchtop thermal slide processer that is used on formalin-fixed, paraffin embedded tissues prior to immunostaining.