missing translation for 'onlineSavingsMsg'
Learn More

(R)-Baclofen 95.0+%, TCI America™

Catalog No. B510225MG
Change view
Click to view available options
Quantity:
25 mg
100 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B510225MG 25 mg
B5102100MG 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B510225MG Supplier TCI America Supplier No. B510225MG
Only null left

Chemical Identifiers

CAS 69308-37-8
Molecular Formula C10H12ClNO2
Molecular Weight (g/mol) 213.66
MDL Number MFCD01321057
InChI Key KPYSYYIEGFHWSV-UHFFFAOYNA-N
Synonym r-baclofen, arbaclofen, r-4-amino-3-4-chlorophenyl butanoic acid, d-baclofen, --baclofen, r---baclofen, 3r-4-amino-3-4-chlorophenyl butanoic acid, unii-nyu6utw25b
PubChem CID 44602
IUPAC Name 4-amino-3-(4-chlorophenyl)butanoic acid
SMILES NCC(CC(O)=O)C1=CC=C(Cl)C=C1

Specifications

Melting Point 176°C
Color White
UN Number 2811
Quantity 25 mg
Formula Weight 213.66
Percent Purity ≥95.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material (R)-Baclofen
RTECSNumber DA8339450
TSCA No