missing translation for 'onlineSavingsMsg'
Learn More

(R)-(-)-alpha-(Trifluoromethyl)benzyl Alcohol, TCI America™

Catalog No. P13671G
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P13671G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. P13671G Supplier TCI America Supplier No. P13671G

Chemical Identifiers

CAS 10531-50-7
Molecular Formula C8H7F3O
Molecular Weight (g/mol) 176.138
MDL Number MFCD00077844
InChI Key VNOMEAQPOMDWSR-SSDOTTSWSA-N
Synonym (R)-(-)-1-Phenyl-2,2,2-trifluoroethanol
PubChem CID 1472402
IUPAC Name (1R)-2,2,2-trifluoro-1-phenylethanol
SMILES C1=CC=C(C=C1)C(C(F)(F)F)O

Specifications

Color Yellow
Boiling Point 99°C
Quantity 1 g
Formula Weight 176.14
Physical Form Liquid
Chemical Name or Material (R)-(-)-alpha-(Trifluoromethyl)benzyl Alcohol