missing translation for 'onlineSavingsMsg'
Learn More

(R)-(+)-alpha-Methylbenzyl Isocyanate 98.0+%, TCI America™

Catalog No. p-7131342
Change view
Click to view available options
Quantity:
1 g
5 g
Catalog No. Quantity
I03341G 1 g
I03345G 5 g
2 options

Catalog No. I03341G

Supplier: TCI America I03341G

Only null left

Chemical Identifiers

CAS 33375-06-3
Molecular Formula C9H9NO
Molecular Weight (g/mol) 147.177
MDL Number MFCD00063015
InChI Key JJSCUXAFAJEQGB-MRVPVSSYSA-N
Synonym r-+-1-phenylethyl isocyanate, 1r-1-isocyanatoethyl benzene, r-+-alpha-methylbenzyl isocyanate, benzene, 1r-1-isocyanatoethyl, isocyanic acid r-+-1-phenylethyl ester, isocyanic acid r-+-alpha-methylbenzyl ester, isocyanic acid-r-+-methylbenzyl ester, 1-isocyanatoethyl benzene #, +-alpha-methylbenzyl isocyanate, pubchem8096
PubChem CID 7018262
IUPAC Name [(1R)-1-isocyanatoethyl]benzene
SMILES CC(C1=CC=CC=C1)N=C=O

Specifications

Color Yellow
Quantity 1 g
UN Number 2206
Formula Weight 147.18
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material (R)-(+)-alpha-Methylbenzyl Isocyanate
TSCA No
Recommended Storage Refrigerator