missing translation for 'onlineSavingsMsg'
Learn More

(R)-(+)-alpha-Hydroxy-gamma-butyrolactone 98.0+%, TCI America™

$689.70 - $3312.43

Chemical Identifiers

CAS 56881-90-4
Molecular Formula C4H6O3
Molecular Weight (g/mol) 102.09
MDL Number MFCD00211246
InChI Key FWIBCWKHNZBDLS-UHFFFAOYNA-N
Synonym (R)-(+)-Dihydro-3-hydroxy-2(3H)-furanone, (R)-(+)-3-Hydroxytetrahydrofuran-2-one
PubChem CID 7157070
IUPAC Name 3-hydroxyoxolan-2-one
SMILES OC1CCOC1=O
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
H0950100MG
View Documents
TCI America
H0950100MG
100 mg
Each for $689.70
Only null left
 
H09501G
View Documents
TCI America
H09501G
1 g
Each for $3,312.43
Only null left
 
Description

Description

Specifications

Chemical Identifiers

56881-90-4
102.09
FWIBCWKHNZBDLS-UHFFFAOYNA-N
7157070
OC1CCOC1=O
C4H6O3
MFCD00211246
(R)-(+)-Dihydro-3-hydroxy-2(3H)-furanone, (R)-(+)-3-Hydroxytetrahydrofuran-2-one
3-hydroxyoxolan-2-one

Specifications

56881-90-4
133°C
MFCD00211246
(R)-(+)-Dihydro-3-hydroxy-2(3H)-furanone, (R)-(+)-3-Hydroxytetrahydrofuran-2-one
OC1CCOC1=O
102.09
102.09
Liquid
Yellow
C4H6O3
100 mg
FWIBCWKHNZBDLS-UHFFFAOYNA-N
3-hydroxyoxolan-2-one
7157070
≥98.0% (GC)
(R)-(+)-alpha-Hydroxy-gamma-butyrolactone
Videos
Safety and Handling

Safety and Handling

TSCA : No

SDS
missing translation for 'documents'

missing translation for 'documents'