missing translation for 'onlineSavingsMsg'
Learn More

(R)-(+)-alpha-Amino-gamma-butyrolactone Hydrochloride 98.0+%, TCI America™

$195.73 - $674.79

Chemical Identifiers

CAS 104347-13-9
Molecular Formula C4H8ClNO2
Molecular Weight (g/mol) 137.563
MDL Number MFCD00674071
InChI Key XBKCXPRYTLOQKS-AENDTGMFSA-N
Synonym d-homoserine lactone hydrochloride, r-+-alpha-amino-gamma-butyrolactone hydrochloride, r-3-aminodihydrofuran-2 3h-one hydrochloride, r-+-alpha-amino-gamma-butyrolactone hydrochloride, r-3-aminodihydrofuran-2 3h-one hydrochloride, 2 3h-furanone, 3-aminodihydro-, hydrochloride, 3r, homoserine lactone hydrochloride, 2 3h-furanone, 3-aminodihydro-, hydrochloride, 3r, r-+-a-amino-g-butyrolactone hydrochloride, pubchem14743, r-+-, a-amino-
PubChem CID 11954435
IUPAC Name (3R)-3-aminooxolan-2-one;hydrochloride
SMILES C1COC(=O)C1N.Cl
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
A25121G
View Documents
TCI America
A25121G
1 g
Each for $195.73
Only null left
 
A25125G
View Documents
TCI America
A25125G
5 g
Each for $674.79
Only null left
 
Description

Description

Specifications

Chemical Identifiers

104347-13-9
137.563
XBKCXPRYTLOQKS-AENDTGMFSA-N
11954435
C1COC(=O)C1N.Cl
C4H8ClNO2
MFCD00674071
d-homoserine lactone hydrochloride, r-+-alpha-amino-gamma-butyrolactone hydrochloride, r-3-aminodihydrofuran-2 3h-one hydrochloride, r-+-alpha-amino-gamma-butyrolactone hydrochloride, r-3-aminodihydrofuran-2 3h-one hydrochloride, 2 3h-furanone, 3-aminodihydro-, hydrochloride, 3r, homoserine lactone hydrochloride, 2 3h-furanone, 3-aminodihydro-, hydrochloride, 3r, r-+-a-amino-g-butyrolactone hydrochloride, pubchem14743, r-+-, a-amino-
(3R)-3-aminooxolan-2-one;hydrochloride

Specifications

104347-13-9
White-Yellow
MFCD00674071
d-homoserine lactone hydrochloride, r-+-alpha-amino-gamma-butyrolactone hydrochloride, d-homoserine lactone hcl, 3r-3-aminotetrahydrofuran-2-one hydrochloride, r-3-aminodihydrofuran-2 3h-one hydrochloride, 2 3h-furanone, 3-aminodihydro-, hydrochloride, 3r, homoserine lactone hydrochloride, 2 3h-furanone, 3-aminodihydro-, hydrochloride, 3r, r-+-, a-amino-, r-+-, a-amino-
C1COC(=O)C1N.Cl
137.563
137.56
Crystalline Powder
223°C
C4H8ClNO2
1 g
XBKCXPRYTLOQKS-AENDTGMFSA-N
(3R)-3-aminooxolan-2-one;hydrochloride
11954435
≥98.0% (N,T)
(R)-(+)-alpha-Amino-gamma-butyrolactone Hydrochloride
Videos
Safety and Handling

Safety and Handling

TSCA : No

SDS
missing translation for 'documents'

missing translation for 'documents'