missing translation for 'onlineSavingsMsg'
Learn More

(R)-(+)-alpha-Amino-gamma-butyrolactone Hydrochloride 98.0+%, TCI America™

Catalog No. A25125G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A25125G 5 g
A25121G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A25125G Supplier TCI America Supplier No. A25125G

Chemical Identifiers

CAS 104347-13-9
Molecular Formula C4H8ClNO2
Molecular Weight (g/mol) 137.563
MDL Number MFCD00674071
InChI Key XBKCXPRYTLOQKS-AENDTGMFSA-N
Synonym d-homoserine lactone hydrochloride, r-+-alpha-amino-gamma-butyrolactone hydrochloride, r-3-aminodihydrofuran-2 3h-one hydrochloride, 3r-3-aminotetrahydrofuran-2-one hydrochloride, homoserine lactone hydrochloride, 2 3h-furanone, 3-aminodihydro-, hydrochloride, 3r, r-+-a-amino-g-butyrolactone hydrochloride, pubchem14743, r-+-, a-amino-, a-butyrolactone hydrochloride, d-homoserine lactone hcl
PubChem CID 11954435
IUPAC Name (3R)-3-aminooxolan-2-one;hydrochloride
SMILES C1COC(=O)C1N.Cl

Specifications

Melting Point 223°C
Color White-Yellow
Quantity 5 g
Formula Weight 137.56
Percent Purity ≥98.0% (N,T)
Physical Form Crystalline Powder
Chemical Name or Material (R)-(+)-alpha-Amino-gamma-butyrolactone Hydrochloride