missing translation for 'onlineSavingsMsg'
Learn More
Learn More
(R)-(-)-alpha-[[(4-Ethyl-2,3-dioxo-1-piperazinyl)carbonyl]amino]benzeneacetic Acid 98.0+%, TCI America™
$52.19 - $72.70
Chemical Identifiers
| CAS | 63422-71-9 |
|---|---|
| Molecular Formula | C15H16N3O5 |
| Molecular Weight (g/mol) | 318.31 |
| MDL Number | MFCD00799536 |
| InChI Key | JQEHQELQPPKXRR-LLVKDONJSA-M |
| Synonym | (R)-(-)-2-(4-Ethyl-2,3-dioxopiperazine-1-carboxamido)-2-phenylacetic Acid |
| PubChem CID | 2733658 |
| IUPAC Name | (2R)-2-[(4-ethyl-2,3-dioxopiperazine-1-carbonyl)amino]-2-phenylacetate |
| SMILES | CCN1CCN(C(=O)N[C@@H](C([O-])=O)C2=CC=CC=C2)C(=O)C1=O |
Chemical Identifiers
| 63422-71-9 | |
| 318.31 | |
| JQEHQELQPPKXRR-LLVKDONJSA-M | |
| 2733658 | |
| CCN1CCN(C(=O)N[C@@H](C([O-])=O)C2=CC=CC=C2)C(=O)C1=O |
| C15H16N3O5 | |
| MFCD00799536 | |
| (R)-(-)-2-(4-Ethyl-2,3-dioxopiperazine-1-carboxamido)-2-phenylacetic Acid | |
| (2R)-2-[(4-ethyl-2,3-dioxopiperazine-1-carbonyl)amino]-2-phenylacetate |
Specifications
| 63422-71-9 | |
| White | |
| MFCD00799536 | |
| (R)-(-)-2-(4-Ethyl-2,3-dioxopiperazine-1-carboxamido)-2-phenylacetic Acid | |
| CCN1CCN(C(=O)N[C@@H](C([O-])=O)C2=CC=CC=C2)C(=O)C1=O | |
| 318.31 | |
| 319.32 | |
| Crystalline Powder |
| 173°C | |
| C15H16N3O5 | |
| 1 g | |
| JQEHQELQPPKXRR-LLVKDONJSA-M | |
| (2R)-2-[(4-ethyl-2,3-dioxopiperazine-1-carbonyl)amino]-2-phenylacetate | |
| 2733658 | |
| ≥98.0% (HPLC,T) | |
| (R)-(-)-alpha-[[(4-Ethyl-2,3-dioxo-1-piperazinyl)carbonyl]amino]benzeneacetic Acid |
Safety and Handling
TSCA : No