missing translation for 'onlineSavingsMsg'
Learn More
Learn More
(R)-(-)-alpha-[[(4-Ethyl-2,3-dioxo-1-piperazinyl)carbonyl]amino]benzeneacetic Acid 98.0+%, TCI America™
Specifications
Specifications
| Melting Point | 173°C |
| Color | White |
| Quantity | 5 g |
| Formula Weight | 319.32 |
| Percent Purity | ≥98.0% (HPLC,T) |
| Physical Form | Crystalline Powder |
| Chemical Name or Material | (R)-(-)-alpha-[[(4-Ethyl-2,3-dioxo-1-piperazinyl)carbonyl]amino]benzeneacetic Acid |
Safety and Handling
| TSCA | No |