missing translation for 'onlineSavingsMsg'
Learn More

(R)-(-)-alpha-[[(4-Ethyl-2,3-dioxo-1-piperazinyl)carbonyl]amino]benzeneacetic Acid 98.0+%, TCI America™

Catalog No. E09931G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
E09931G 1 g
E09935G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. E09931G Supplier TCI America Supplier No. E09931G
Only null left

Chemical Identifiers

CAS 63422-71-9
Molecular Formula C15H16N3O5
Molecular Weight (g/mol) 318.31
MDL Number MFCD00799536
InChI Key JQEHQELQPPKXRR-LLVKDONJSA-M
Synonym (R)-(-)-2-(4-Ethyl-2,3-dioxopiperazine-1-carboxamido)-2-phenylacetic Acid
PubChem CID 2733658
IUPAC Name (2R)-2-[(4-ethyl-2,3-dioxopiperazine-1-carbonyl)amino]-2-phenylacetate
SMILES CCN1CCN(C(=O)N[C@@H](C([O-])=O)C2=CC=CC=C2)C(=O)C1=O

Specifications

Melting Point 173°C
Color White
Quantity 1 g
Formula Weight 319.32
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material (R)-(-)-alpha-[[(4-Ethyl-2,3-dioxo-1-piperazinyl)carbonyl]amino]benzeneacetic Acid