missing translation for 'onlineSavingsMsg'
Learn More

(R)-(+)-alpha-(1-Naphthyl)ethylamine, 99+%

Catalog No. p-170717
Change view
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC217800010 1 g Glass bottle
AC217800050 5 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC217800010 Supplier Thermo Scientific Chemicals Supplier No. 217800010

CAS: 3886-70-2 | C12H13N | 171.24 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chiral derivatization reagent

Chemical Identifiers

CAS 3886-70-2
Molecular Formula C12H13N
Molecular Weight (g/mol) 171.24
MDL Number MFCD00064114
InChI Key RTCUCQWIICFPOD-SECBINFHSA-N
Synonym r-+-1-1-naphthyl ethylamine, r-1-naphthalen-1-yl ethanamine, 1r-1-naphthalen-1-yl ethan-1-amine, r-1-1-naphthyl ethylamine, 1r-1-naphthalen-1-ylethanamine, 1r-1-naphthylethylamine, r-+-alpha-1-naphthyl ethylamine, 1r-1-1-naphthyl ethylamine, r-+-a-1-naphthyl ethylamine, 1r-1-naphthalen-1-yl ethanamine
PubChem CID 2724264
IUPAC Name (1R)-1-naphthalen-1-ylethanamine
SMILES CC(C1=CC=CC2=CC=CC=C21)N

Specifications

CAS Min % 99.0
CAS Max % 100.0
Color Yellow
Density 1.0600g/mL
Boiling Point 153.0°C (11.0 mmHg)
Flash Point >110°C
Infrared Spectrum Authentic
Assay Percent Range 99% min. (GC)
Packaging Glass bottle
Refractive Index 1.6217 to 1.6237
Linear Formula C10H7CH(CH3)NH2
Quantity 1 g
Beilstein 12, III, 3110
Specific Gravity 1.06
Specific Rotation Condition + 60.00 (20.00°C c=2,CH3OH)
Specific Rotation + 60.00
Formula Weight 171.24
Percent Purity 99+%
Physical Form Liquid
Chemical Name or Material (R)-(+)-α-(1-Naphthyl)ethylamine, 99+%
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber 223-425-5
TSCA TSCA

RUO – Research Use Only