missing translation for 'onlineSavingsMsg'
Learn More

(R)-6,6'-Dibromo-2,2'-bis(methoxymethoxy)-1,1'-binaphthyl 98.0+%, TCI America™

Catalog No. p-7128523
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D48821G 1 g
D48825G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D48821G Supplier TCI America Supplier No. D48821G
Only null left

Chemical Identifiers

CAS 179866-74-1
Molecular Formula C24H20Br2O4
Molecular Weight (g/mol) 532.23
MDL Number MFCD03788936
InChI Key UPLLZDVWXACTEG-UHFFFAOYSA-N
PubChem CID 10839989
IUPAC Name 6,6'-dibromo-2,2'-bis(methoxymethoxy)-1,1'-binaphthalene
SMILES COCOC1=CC=C2C=C(Br)C=CC2=C1C1=C2C=CC(Br)=CC2=CC=C1OCOC

Specifications

Melting Point 135°C
Color White-Yellow
Quantity 1 g
Formula Weight 532.23
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material (R)-6,6′-Dibromo-2,2′-bis(methoxymethoxy)-1,1′-binaphthyl