missing translation for 'onlineSavingsMsg'
Learn More

(R)-(-)-5-Oxotetrahydrofuran-2-carboxylic Acid 98.0+%, TCI America™

Catalog No. p-7134274
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
O02811G 1 g
O02815G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. O02811G Supplier TCI America Supplier No. O02811G

Chemical Identifiers

CAS 53558-93-3
Molecular Formula C5H6O4
Molecular Weight (g/mol) 130.099
MDL Number MFCD00011567
InChI Key QVADRSWDTZDDGR-GSVOUGTGSA-N
Synonym r-5-oxotetrahydrofuran-2-carboxylic acid, r---5-oxotetrahydrofuran-2-carboxylic acid, 2r-5-oxooxolane-2-carboxylic acid, r---5-oxotetrahydrofuran-2-carboxylicacid, 2-furancarboxylic acid, tetrahydro-5-oxo-, 2r, r-5-oxotetrahydrofuran-2-carboxylicacid, r---5-oxo-2-tetrahydrofurancarboxylic acid, r-5-oxotetrahydro-2-furancarboxylic acid, 5-oxo-tetrahydro-furan-2-carboxylic, 5-furancarboxylic acid, tetrahydro-2-oxo-, r--
PubChem CID 104532
IUPAC Name (2R)-5-oxooxolane-2-carboxylic acid
SMILES C1CC(=O)OC1C(=O)O

Specifications

Melting Point 72°C
Color White-Yellow
Quantity 1 g
Formula Weight 130.10
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material (R)-(-)-5-Oxotetrahydrofuran-2-carboxylic Acid