missing translation for 'onlineSavingsMsg'
Learn More

(R)-5-Bromo-3-[1-(2,6-dichloro-3-fluorophenyl)ethoxy]pyridin-2-amine 98.0+%, TCI America™

Catalog No. B56821G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B56821G 1 g
B56825G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B56821G Supplier TCI America Supplier No. B56821G
Only null left

Chemical Identifiers

CAS 877399-00-3
Molecular Formula C13H10BrCl2FN2O
Molecular Weight (g/mol) 380.038
MDL Number MFCD18207061
InChI Key URFUZAZEKBBCEY-ZCFIWIBFSA-N
PubChem CID 11689426
IUPAC Name 5-bromo-3-[(1R)-1-(2,6-dichloro-3-fluorophenyl)ethoxy]pyridin-2-amine
SMILES CC(C1=C(C=CC(=C1Cl)F)Cl)OC2=CC(=CN=C2N)Br

Specifications

Melting Point 109°C
Color White-Yellow
Quantity 1 g
Formula Weight 380.04
Percent Purity ≥98.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material (R)-5-Bromo-3-[1-(2,6-dichloro-3-fluorophenyl)ethoxy]pyridin-2-amine