missing translation for 'onlineSavingsMsg'
Learn More

(R)-(-)-4-Penten-2-ol, 95%

Catalog No. AAB2565903
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2565903 1 g
AAB2565906 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAB2565903 Supplier Thermo Scientific Chemicals Supplier No. B2565903
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 64584-92-5 | C5H10O | 86.13 g/mol

(R)-(-)-4-Penten-2-ol is used as a pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
(R)-(-)-4-Penten-2-ol is used as a pharmaceutical intermediate.

Solubility
Soluble in water, 4.526 mg/L @ 25°C.

Notes
Store in cool, dry place in a well sealed container. Store away from oxidizing agents.

Chemical Identifiers

CAS 64584-92-5
Molecular Formula C5H10O
Molecular Weight (g/mol) 86.13
MDL Number MFCD03701536
InChI Key ZHZCYWWNFQUZOR-UHFFFAOYNA-N
Synonym r---4-penten-2-ol, 2r-pent-4-en-2-ol, r-pent-4-en-2-ol, 4-penten-2-ol, 2r, r-4-penten-2-ol, 2r-4-penten-2-ol, 4-penten-2 r-ol, r-2-hydroxy-4-pentene
PubChem CID 6999724
SMILES CC(O)CC=C

Specifications

Density 0.837
Boiling Point 115°C to 116°C
Flash Point 30°C (86°F)
Refractive Index 1.424
Quantity 1 g
UN Number UN1987
Solubility Information Soluble in water,4.526mg/L @ 25°C.
Optical Rotation −5.2° (c=1 in Chloroform)
IUPAC Name pent-4-en-2-ol
Formula Weight 86.13
Percent Purity 95%
Chemical Name or Material (R)-(-)-4-Penten-2-ol
Show More Show Less
Hazard Category H226
Hazard Statement GHS H Statement
H226
Flammable liquid and vapour.
Precautionary Statement P210-P233-P240-P241-P242-P243-P280-P303+P361+P353-P370+P378q-P501c
DOTInformation Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: ALCOHOLS, N.O.S.
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only