missing translation for 'onlineSavingsMsg'
Learn More

(R)-(-)-4-Methylmandelic acid, ChiPros∣r, 98%, ee 97+%

Catalog No. p-7028566
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH2595303 1 g
AAH2595306 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH2595303 Supplier Thermo Scientific Chemicals Supplier No. H2595303

CAS: 31284-89-6 | C9H10O3 | 166.18 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 31284-89-6
Molecular Formula C9H10O3
Molecular Weight (g/mol) 166.18
MDL Number MFCD00798433,MFCD00961860
InChI Key SFGURAWGCAPHON-UHFFFAOYNA-N
Synonym 2-hydroxy-2-4-methylphenyl acetic acid, 4-methylmandelic acid, p-methyl mandelic acid, 2-hydroxy-2-p-tolyl acetic acid, hydroxy 4-methylphenyl acetic acid, hydroxy-p-tolyl-acetic acid, p-methylmandelic acid, 4-methylmaldelic acid, p-methyl-mandelic acid, acmc-1afsj
PubChem CID 12458823
IUPAC Name (2R)-2-hydroxy-2-(4-methylphenyl)acetic acid
SMILES CC1=CC=C(C=C1)C(O)C(O)=O

Specifications

Melting Point 130°C to 133°C
Quantity 1 g
Sensitivity Light sensitive
Optical Rotation −154° (c=1 in Methanol)
Formula Weight 166.18
Percent Purity 98%
Assay ee ≥97%
Chemical Name or Material (R)-(-)-4-Methylmandelic acid, ChiPros
Hazard Category H315-H318-H335
Hazard Statement GHS H Statement
H318-H335-H315
Causes serious eye damage.
May cause respiratory irritation.
Causes skin irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P310-P312-P332+P313-P362-P501c
EINECSNumber 662-983-8
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only