Learn More
(R)-(-)-4-Methyl-2-pentanol, 99%
CAS: 16404-54-9 | C6H14O | 102.177 g/mol
Supplier: Thermo Scientific Chemicals L1888103
Description
For synthesis of optically active products
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
| (R)-(-)-4-Methyl-2-pentanol | |
| 0.802 | |
| 41°C (105°F) | |
| C6H14O | |
| MFCD03093077 | |
| UN2053 | |
| r---4-methyl-2-pentanol, 2r-4-methylpentan-2-ol, r-4-methyl-2-pentanol, r-4-methylpentan-2-ol, r---4-methylpentan-2-ol, 4-methyl r-2-pentanol, 2r-4-methyl-2-pentanol, r---methylpentan-2-ol, r--4-methyl-2-pentanol, 2-pentanol, 4-methyl-, 2r | |
| −21° (Neat) | |
| (2R)-4-methylpentan-2-ol | |
| 6999987 | |
| 99% |
| 16404-54-9 | |
| 130°C to 131°C | |
| Slightly Sweetish | |
| 1.411 | |
| 1 g | |
| 1718992 | |
| WVYWICLMDOOCFB-ZCFIWIBFSA-N | |
| CC(C)CC(C)O | |
| 102.177 | |
| 102.18 |
Chemical Identifiers
| 16404-54-9 | |
| 102.177 | |
| WVYWICLMDOOCFB-ZCFIWIBFSA-N | |
| 6999987 | |
| CC(C)CC(C)O |
| C6H14O | |
| MFCD03093077 | |
| r---4-methyl-2-pentanol, 2r-4-methylpentan-2-ol, r-4-methyl-2-pentanol, r-4-methylpentan-2-ol, r---4-methylpentan-2-ol, 4-methyl r-2-pentanol, 2r-4-methyl-2-pentanol, r---methylpentan-2-ol, r--4-methyl-2-pentanol, 2-pentanol, 4-methyl-, 2r | |
| (2R)-4-methylpentan-2-ol |
Safety and Handling
GHS H Statement
H226-H335
Flammable liquid and vapor.
May cause respiratory irritation.
P210-P233-P235-P240-P241-P242-P243-P261-P271-P280-P303+P361+P353-P304+P340-P312-P370+P378q-P501c
H226-H332-H335
DOTInformation : Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: METHYL ISOBUTYL CARBINOL
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only