missing translation for 'onlineSavingsMsg'
Learn More

(R)-(-)-4-Isopropyl-3-propionyl-2-oxazolidinone 98.0+%, TCI America™

Catalog No. I05945G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
I05945G 5 g
I05941G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. I05945G Supplier TCI America Supplier No. I05945G
Only null left

Chemical Identifiers

CAS 89028-40-0
Molecular Formula C9H15NO3
Molecular Weight (g/mol) 185.223
MDL Number MFCD00269691
InChI Key HOWPHXVPNNPSAZ-ZETCQYMHSA-N
Synonym (R)-(-)-3-Propionyl-4-isopropyl-2-oxazolidinone
PubChem CID 643512
IUPAC Name (4R)-3-propanoyl-4-propan-2-yl-1,3-oxazolidin-2-one
SMILES CCC(=O)N1C(COC1=O)C(C)C

Specifications

Color Yellow
Quantity 5 g
Formula Weight 185.22
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material (R)-(-)-4-Isopropyl-3-propionyl-2-oxazolidinone