missing translation for 'onlineSavingsMsg'
Learn More

(R)-4-Boc-morpholine-3-carboxylic acid, 97%

Catalog No. p-7055937
Change view
Click to view available options
Quantity:
250 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH64654MD 250 mg
AAH6465403 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH64654MD Supplier Thermo Scientific Chemicals Supplier No. H64654MD
Only null left

CAS: 869681-70-9 | C10H17NO5 | 231.248 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 869681-70-9
Molecular Formula C10H17NO5
Molecular Weight (g/mol) 231.248
MDL Number MFCD04114909
InChI Key KVXXEKIGMOEPSA-SSDOTTSWSA-N
Synonym r-4-tert-butoxycarbonyl morpholine-3-carboxylic acid, r-4-boc-morpholine-3-carboxylic acid, 4-boc-3 r-morpholinecarboxylic acid, r-morpholine-3,4-dicarboxylic acid 4-tert-butyl ester, r-n-boc-morpholine-3-carboxylic acid, r-4-n-boc-3-morpholinecarboxylic acid, r-4-boc-morpholine-3-carboxylicacid, 3r-4-tert-butoxycarbonyl morpholine-3-carboxylic acid, 3,4-morpholinedicarboxylic acid, 4-1,1-dimethylethyl ester, 3r, ambotzbaa5270
PubChem CID 1512538
IUPAC Name (3R)-4-[(2-methylpropan-2-yl)oxycarbonyl]morpholine-3-carboxylic acid
SMILES CC(C)(C)OC(=O)N1CCOCC1C(=O)O

Specifications

Melting Point 181°C
Color White
UN Number UN2811
Beilstein 10254221
Quantity 250 mg
Formula Weight 231.25
Percent Purity 97%
Physical Form Powder
Chemical Name or Material (R)-4-Boc-morpholine-3-carboxylic acid
Hazard Category H302+H312+H332-H315-H319-H335
Hazard Statement GHS H Statement
H302-H312-H332-H315-H319-H335
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
DOTInformation Transport Hazard Class: 6.1; Packaging Group: III; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.
TSCA No
storageNote1 -30° to -10°C
Recommended Storage Store at -20°C

RUO – Research Use Only