missing translation for 'onlineSavingsMsg'
Learn More

(R)-(-)-4-Benzyl-3-propionyl-2-oxazolidinone 98.0+%, TCI America™

Catalog No. B21651G
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B21651G 1 g
B21655G 5 g
B216525G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. B21651G Supplier TCI America Supplier No. B21651G
Only null left

Chemical Identifiers

CAS 131685-53-5
Molecular Formula C13H15NO3
Molecular Weight (g/mol) 233.267
MDL Number MFCD00269687
InChI Key WHOBYFHKONUTMW-LLVKDONJSA-N
PubChem CID 10966403
IUPAC Name (4R)-4-benzyl-3-propanoyl-1,3-oxazolidin-2-one
SMILES CCC(=O)N1C(COC1=O)CC2=CC=CC=C2

Specifications

Melting Point 45°C
Color White-Yellow
Quantity 1 g
Formula Weight 233.27
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material (R)-(-)-4-Benzyl-3-propionyl-2-oxazolidinone