missing translation for 'onlineSavingsMsg'
Learn More

(R)-4-(4-Methoxybenzyloxy)-1,2-butanediol 96.0+%, TCI America™

Catalog No. M1788500MG
Change view
Click to view available options
Quantity:
500 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M1788500MG 500 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. M1788500MG Supplier TCI America Supplier No. M1788500MG
Only null left

Chemical Identifiers

CAS 213978-61-1
Molecular Formula C12H18O4
Molecular Weight (g/mol) 226.272
InChI Key YRHIJTKHDXQBLC-LLVKDONJSA-N
PubChem CID 44629947
IUPAC Name (2R)-4-[(4-methoxyphenyl)methoxy]butane-1,2-diol
SMILES COC1=CC=C(C=C1)COCCC(CO)O

Specifications

Quantity 500 mg
Formula Weight 226.27
Percent Purity ≥96.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material (R)-4-(4-Methoxybenzyloxy)-1,2-butanediol