Learn More
BUCHI Rotavapor R-300 Rotary Evaporators
Catalog No. 05000977
Change view
Click to view available options
Condenser Type:
Cold Trap
Vertical
Vertical Coil for cold water or a chiller
Description:
Electronic lift
Manual lift
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Condenser Type Description
05000977 Vertical Electronic lift
05000491 Vertical Manual lift
05000485 Vertical Coil for cold water or a chiller Manual lift
05000947 Cold Trap Electronic lift
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 05000977 Supplier BUCHI Supplier No. 11R300252V102
  • ONLINE EXCLUSIVE PRICE
    $15,527.96
    $9,123.56 / Each
    Save $6,404.40
    41.0% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
View all promotions
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
The R-300 meets the highest expectations in convenience and versatility. Its modular design allows extension of the Rotavapor R-300 to a fully integrated system where a central interface regulates each component.
  • Intuitive user interaction
  • Precise operation control for repeatability and safety
  • 1 L or 5 L flask capacity baths with cordless changes and cleaning
  • Up to 220 °C bath heating with 5 L bath
  • Ergonomic handle with manual or electronic lift
  • Choice of P+G coating for added protection of condenser and receiving flask
  • Unattended operation and mobile monitoring available
  • Modular system for control, vacuum, and chiller integration
  • Additional glass assemblies and configurations
  • Cloud services for optimized productivity and automated processes
Certifications
CE and CSA certified
Specifications
Description Electronic lift
Voltage 100/120 V
Capacity (Metric) 5 L
Speed 10 to 280 rpm
Temperature Range (Metric) 20°C to 220°C
For Use With (Application) Standard Distillation (for use with cold water or a chiller)
Condenser Type Vertical
Certifications CE / CSA
Type Rotary Evaporator
Frequency 50/60 Hz
Model R-300
Standard Taper Joint 24/40
Show More Show Less