Learn More
BUCHI Rotavapor R-300 Rotary Evaporators
Catalog No. 05000491
Change view
Click to view available options
Condenser Type:
Cold Trap
Vertical
Vertical Coil for cold water or a chiller
Description:
Electronic lift
Manual lift
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Condenser Type Description
05000491 Vertical Manual lift
05000485 Vertical Coil for cold water or a chiller Manual lift
05000947 Cold Trap Electronic lift
05000977 Vertical Electronic lift
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 05000491 Supplier BUCHI Supplier No. 11R300152V102
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
The R-300 meets the highest expectations in convenience and versatility. Its modular design allows extension of the Rotavapor R-300 to a fully integrated system where a central interface regulates each component.
  • Intuitive user interaction
  • Precise operation control for repeatability and safety
  • 1 L or 5 L flask capacity baths with cordless changes and cleaning
  • Up to 220 °C bath heating with 5 L bath
  • Ergonomic handle with manual or electronic lift
  • Choice of P+G coating for added protection of condenser and receiving flask
  • Unattended operation and mobile monitoring available
  • Modular system for control, vacuum, and chiller integration
  • Additional glass assemblies and configurations
  • Cloud services for optimized productivity and automated processes
Certifications
CE and CSA certified
Specifications
Description Manual lift
Voltage 100/120 V
Capacity (Metric) 5 L
Speed 10 to 280 rpm
Temperature Range (Metric) 20°C to 220°C
For Use With (Application) Standard Distillation (for use with cold water or a chiller)
Condenser Type Vertical
Certifications CE / CSA
Type Rotary Evaporator
Frequency 50/60 Hz
Model R-300
Standard Taper Joint 24/40
Show More Show Less