missing translation for 'onlineSavingsMsg'
Learn More

(R)-(+)-3-(tert-Butoxycarbonyl)-4-methoxycarbonyl-2,2-dimethyl-1,3-oxazolidine 96.0+%, TCI America™

Catalog No. B53525G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B53525G 5 g
B53521G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B53525G Supplier TCI America Supplier No. B53525G

Chemical Identifiers

CAS 95715-86-9
Molecular Formula C12H21NO5
Molecular Weight (g/mol) 259.30
MDL Number MFCD00674041
InChI Key ZNBUXTFASGDVCL-UHFFFAOYNA-N
Synonym r-3-tert-butyl 4-methyl 2,2-dimethyloxazolidine-3,4-dicarboxylate, methyl r-+-3-tert-butoxycarbonyl-2,2-dimethyl-4-oxazolidinecarboxylate, 3-tert-butyl 4-methyl 4r-2,2-dimethyl-1,3-oxazolidine-3,4-dicarboxylate, r-+-3-tert-butoxycarbonyl-2,2-dimethyl-4-oxazolidinecarboxylic acid methyl ester, methyl r-n-boc-2,2-dimethyloxazolidine-4-carboxylate, r-+-3-boc-4-methoxycarbonyl-2,2-dimethyl-1,3-oxazolidine, r---3-boc-4-methoxycarbonyl-2,2-dimethyl-1,3-oxazolidine, methyl r-+-3-boc-2,2-dimethyl-4-oxazolidinecarboxylate, 3-tert-butyl 4-methyl r-+-2,2-dimethyloxazolidine-3,4-dicarboxylate, methyl 3-tert-butoxycarbonyl-2,2-dimethyloxazolidine-4-r-carboxylate
PubChem CID 688220
IUPAC Name 3-tert-butyl 4-methyl 2,2-dimethyl-1,3-oxazolidine-3,4-dicarboxylate
SMILES COC(=O)C1COC(C)(C)N1C(=O)OC(C)(C)C

Specifications

Color Yellow
Boiling Point 102°C
Quantity 5 g
Formula Weight 259.30
Percent Purity ≥96.0% (GC)
Physical Form Liquid
Chemical Name or Material (R)-(+)-3-(tert-Butoxycarbonyl)-4-methoxycarbonyl-2,2-dimethyl-1,3-oxazolidine