missing translation for 'onlineSavingsMsg'
Learn More

(R)-(-)-3-Pyrrolidinol Hydrochloride 98.0+%, TCI America™

Catalog No. p-7134970
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P11405G 5 g
P114025G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. P11405G Supplier TCI America Supplier No. P11405G
Only null left

Chemical Identifiers

CAS 104706-47-0
Molecular Formula C4H10ClNO
Molecular Weight (g/mol) 123.58
MDL Number MFCD00191570
InChI Key QPMSJEFZULFYTB-PGMHMLKASA-N
Synonym r-3-hydroxypyrrolidine hydrochloride, r---3-pyrrolidinol hydrochloride, 3r-pyrrolidin-3-ol hydrochloride, r-pyrrolidin-3-ol hydrochloride, r-3-hydroxypyrrolidine hcl, r-3-pyrrolidinol hydrochloride, 3r-3-hydroxypyrrolidine hydrochloride, 3r-pyrrolidinol hydrochloride, r-+-3-pyrrolidinol hydrochloride
PubChem CID 2759336
IUPAC Name (3R)-pyrrolidin-3-ol;hydrochloride
SMILES C1CNCC1O.Cl

Specifications

Melting Point 109°C
Quantity 5 g
Formula Weight 123.58
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material (R)-(-)-3-Pyrrolidinol Hydrochloride