missing translation for 'onlineSavingsMsg'
Learn More

(R)-(+)-3-Pyrrolidinol, 98%

Catalog No. p-189920
Change view
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC344570010 1 g Glass bottle
AC344570050 5 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC344570010 Supplier Thermo Scientific Chemicals Supplier No. 344570010
Only null left

CAS: 2799-21-5 | C4H9NO | 87.12 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 2799-21-5
Molecular Formula C4H9NO
Molecular Weight (g/mol) 87.12
MDL Number MFCD00145220
InChI Key JHHZLHWJQPUNKB-SCSAIBSYSA-N
Synonym r-3-hydroxypyrrolidine, r-pyrrolidin-3-ol, r-3-pyrrolidinol, 3r-pyrrolidin-3-ol, 3r-3-pyrrolidinol, r-hydroxypyrrolidine, r-+-3-hydroxypyrrolidine, r-hp, 3-pyrrolidinol, 3r, 3r-pyrrolidinol
PubChem CID 2759337
IUPAC Name (3R)-pyrrolidin-3-ol
SMILES C1CNCC1O

Specifications

Density 1.0480g/mL
Boiling Point 108°C to 110°C (8.0 mmHg)
Flash Point >110°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Refractive Index 1.4870 to 1.491
Quantity 1 g
Specific Gravity 1.048
Specific Rotation Condition +6° (20°C C=3.5,CH3OH)
Specific Rotation + 6
Formula Weight 87.12
Percent Purity 98%
Physical Form Viscous Liquid
Chemical Name or Material (R)-(+)-3-Pyrrolidinol
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement:
Causes skin irritation.
May cause respiratory irritation.
Causes serious eye irritation.
Precautionary Statement GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Recommended Storage Product darkens on storage

RUO – Research Use Only