missing translation for 'onlineSavingsMsg'
Learn More

(R)-(-)-3-Piperidinecarboxylic Acid 98.0+%, TCI America™

Catalog No. N06545G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
N06545G 5 g
N06541G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. N06545G Supplier TCI America Supplier No. N06545G

Chemical Identifiers

CAS 25137-00-2
Molecular Formula C6H11NO2
Molecular Weight (g/mol) 129.159
MDL Number MFCD01630787
InChI Key XJLSEXAGTJCILF-RXMQYKEDSA-N
Synonym r-piperidine-3-carboxylic acid, r---nipecotic acid, r-nipecotic acid, 3r-piperidine-3-carboxylic acid, r---3-piperidinecarboxylic acid, --nipecotic acid, d--nipecotic acid, 3r---piperidine-3-carboxylic acid, r-3-piperidinecarboxylic acid, 3r-nipecotic acid
PubChem CID 1796510
ChEBI CHEBI:221278
IUPAC Name (3R)-piperidine-3-carboxylic acid
SMILES C1CC(CNC1)C(=O)O

Specifications

Color White
Quantity 5 g
Formula Weight 129.16
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material (R)-(-)-3-Piperidinecarboxylic Acid